Merck
  • Ectodysplasin-A2 induces dickkopf 1 expression in human balding dermal papilla cells overexpressing the ectodysplasin A2 receptor.
Ectodysplasin-A2 induces dickkopf 1 expression in human balding dermal papilla cells overexpressing the ectodysplasin A2 receptor.
Biochemical and biophysical research communications (2020-08-02)
Mi Hee Kwack, Mee Sook Jun, Young Kwan Sung, Jung Chul Kim, Moon Kyu Kim
ZUSAMMENFASSUNG
Androgenetic alopecia (AGA) is a common genetic disorder, and a X-chromosomal locus that contains the androgen receptor (AR) and ectodysplasin A2 receptor (EDA2R) genes represents a major susceptibility locus for AGA. In our previous study, we reported that ectodysplasin-A2 (EDA-A2) induces apoptosis in cultured human hair follicle (HF) cells and promotes the regression of HFs in mice. However, the role of the EDA-A2/EDA2R in AGA remains unknown, as the causative gene in this pathway has not yet been identified and potential functional connections between EDA-A2 signaling and the androgen pathway remain unclear. In this study, we investigated the expression of EDA2R in balding HFs and matched with non-balding HFs. The EDA2R level was upregulated in the balding dermal papilla (DP) cells compared with non-balding DP cells derived from patients with AGA. However, EDA2R was strongly expressed in both balding and non-balding outer root sheath (ORS) cells. We screened EDA-A2-regulated genes in balding DP cells and identified dickkopf 1 (DKK-1) as catagen inducer during the hair cycle. The mRNA and protein expression levels of DKK-1 were both upregulated by EDA-A2. In addition, DKK-1 expression was induced by EDA-A2 both in cultured human HFs and in mouse HFs. Moreover, the EDA-A2-induced apoptosis of DP and ORS cells was reversed by the antibody-mediated neutralization of DKK-1. Collectively, our data strongly suggest that EDA-A2 induces DKK-1 secretion and causes apoptosis in HFs by binding EDA2R, which is overexpressed in the bald scalp. EDA-A2/EDA2R signaling could inhibit hair growth through DKK-1 induction, and an inhibitor of EDA-A2/EDA2R signaling may be a promising agent for the treatment and prevention of AGA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human EDA2R