Merck
  • The role of immune and oxidative pathways in menstrual cycle associated depressive, physio-somatic, breast and anxiety symptoms: Modulation by sex hormones.
The role of immune and oxidative pathways in menstrual cycle associated depressive, physio-somatic, breast and anxiety symptoms: Modulation by sex hormones.
Journal of psychosomatic research (2020-06-12)
Chutima Roomruangwong, Andressa Keiko Matsumoto, Ana Paula Michelin, Laura de Oliveira Semeão, João Victor de Lima Pedrão, Estefania G Moreira, Sunee Sirivichayakul, Andre Carvalho, Decio S Barbosa, Michael Maes
ZUSAMMENFASSUNG
To examine whether 1) immune and nitro-oxidative stress (IO&NS) biomarkers are associated with premenstrual syndrome (PMS); and 2) changes in IO&NS biomarkers during the menstrual cycle (MC) are associated with PMS symptoms and plasma estradiol and progesterone. This longitudinal study examined 41 women who completed the Daily Record of Severity of Problems (DRSP) rating scale during 28 consecutive days and assayed plasma levels of complement C3 and C4, highly sensitive C-reactive protein (hsCRP), haptoglobin (Hp), advanced oxidation protein products (AOPP), lipid hydroperoxides (LOOH), nitric oxide metabolites (NOx), total radical-trapping antioxidant parameter (TRAP), sulfhydryl (-SH) groups and the activity of paraoxonase (PON)1 at days 7 (D7), 14 (D14), 21 (D21) and 28 (D28) of the MC. MC Associated Syndrome (MCAS) was diagnosed when the summed DRSP score during the MC is >0.666 percentile. All biomarkers, except hsCRP, showed significant alterations during the MC. Arylesterase (AREase) was lowered at D28, while LOOH increased at D14 and C4 at D21 in MCAS. Total DRSP scores were predicted by the combined effects of C4 (positively) and AREase and malondialdehyde (MDA) (both inversely associated). Progesterone lowered levels of LOOH, AOPP and C3 and estradiol lowered levels of Hp while both sex hormones increased 4-(chloromethyl)phenyl acetate (CMPA)ase and AREase activities and levels of -SH groups. PMS/MCAS is not accompanied by a peripheral inflammatory response. Lowered MDA and antioxidant defenses and increased C4 may play a role in MC symptoms while sex hormones may have a protective effect against oxidative stress toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(Chlormethyl)phenylacetat, 98%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)