Merck
  • Histidine-Rich Glycoprotein Inhibits High-Mobility Group Box-1-Mediated Pathways in Vascular Endothelial Cells through CLEC-1A.
Histidine-Rich Glycoprotein Inhibits High-Mobility Group Box-1-Mediated Pathways in Vascular Endothelial Cells through CLEC-1A.
iScience (2020-06-05)
Shangze Gao, Hidenori Wake, Masakiyo Sakaguchi, Dengli Wang, Youhei Takahashi, Kiyoshi Teshigawara, Hui Zhong, Shuji Mori, Keyue Liu, Hideo Takahashi, Masahiro Nishibori
ZUSAMMENFASSUNG
High-mobility group box-1 (HMGB1) protein has been postulated to play a pathogenic role in severe sepsis. Histidine-rich glycoprotein (HRG), a 75 kDa plasma protein, was demonstrated to improve the survival rate of septic mice through the regulation of neutrophils and endothelium barrier function. As the relationship of HRG and HMGB1 remains poorly understood, we investigated the effects of HRG on HMGB1-mediated pathway in endothelial cells, focusing on the involvement of specific receptors for HRG. HRG potently inhibited the HMGB1 mobilization and effectively suppressed rHMGB1-induced inflammatory responses and expression of all three HMGB1 receptors in endothelial cells. Moreover, we first clarified that these protective effects of HRG on endothelial cells were mediated through C-type lectin domain family 1 member A (CLEC-1A) receptor. Thus, current study elucidates protective effects of HRG on vascular endothelial cells through inhibition of HMGB1-mediated pathways may contribute to the therapeutic effects of HRG on severe sepsis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium – hoher Glucosegehalt, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli