Merck
  • Multifunctional Biomaterials: Combining Material Modification Strategies for Engineering of Cell-Contacting Surfaces.
Multifunctional Biomaterials: Combining Material Modification Strategies for Engineering of Cell-Contacting Surfaces.
ACS applied materials & interfaces (2020-04-15)
Anne-Sophie Mertgen, Vanessa Tanja Trossmann, Anne Géraldine Guex, Katharina Maniura-Weber, Thomas Scheibel, Markus Rottmar
ZUSAMMENFASSUNG
In the human body, cells in a tissue are exposed to signals derived from their specific extracellular matrix (ECM), such as architectural structure, mechanical properties, and chemical composition (proteins, growth factors). Research on biomaterials in tissue engineering and regenerative medicine aims to recreate such stimuli using engineered materials to induce a specific response of cells at the interface. Although traditional biomaterials design has been mostly limited to varying individual signals, increasing interest has arisen on combining several features in recent years to improve the mimicry of extracellular matrix properties. Tremendous progress in combinatorial surface modification exploiting, for example, topographical features or variations in mechanics combined with biochemical cues has enabled the identification of their key regulatory characteristics on various cell fate decisions. Gradients especially facilitated such research by enabling the investigation of combined continuous changes of different signals. Despite unravelling important synergies for cellular responses, challenges arise in terms of fabrication and characterization of multifunctional engineered materials. This review summarizes recent work on combinatorial surface modifications that aim to control biological responses. Modification and characterization methods for enhanced control over multifunctional material properties are highlighted and discussed. Thereby, this review deepens the understanding and knowledge of biomimetic combinatorial material modification, their challenges but especially their potential.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2-Dimethoxy-2-phenylacetophenon, 99%