Merck
  • Activation of AMP-activated protein kinase during sepsis/inflammation improves survival by preserving cellular metabolic fitness.
Activation of AMP-activated protein kinase during sepsis/inflammation improves survival by preserving cellular metabolic fitness.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-04-05)
Kui Jin, Yujie Ma, Carlos L Manrique-Caballero, Hui Li, David R Emlet, Shengnan Li, Catherine J Baty, Xiaoyan Wen, Nahmah Kim-Campbell, Alicia Frank, Elizabeth V Menchikova, Nuria M Pastor-Soler, Kenneth R Hallows, Edwin K Jackson, Sruti Shiva, Michael R Pinsky, Brian S Zuckerbraun, John A Kellum, Hernando Gómez
ZUSAMMENFASSUNG
The purpose was to determine the role of AMPK activation in the renal metabolic response to sepsis, the development of sepsis-induced acute kidney injury (AKI) and on survival. In a prospective experimental study, 167 10- to 12-week-old C57BL/6 mice underwent cecal ligation and puncture (CLP) and human proximal tubule epithelial cells (TEC; HK2) were exposed to inflammatory mix (IM), a combination of lipopolysaccharide (LPS) and high mobility group box 1 (HMGB1). Renal/TEC metabolic fitness was assessed by monitoring the expression of drivers of oxidative phosphorylation (OXPHOS), the rates of utilization of OXPHOS/glycolysis in response to metabolic stress, and mitochondrial function by measuring O2 consumption rates (OCR) and the membrane potential (Δψm ). Sepsis/IM resulted in AKI, increased mortality, and in renal AMPK activation 6-24 hours after CLP/IM. Pharmacologic activation of AMPK with 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR) or metformin during sepsis improved the survival, while AMPK inhibition with Compound C increased mortality, impaired mitochondrial respiration, decreased OCR, and disrupted TEC metabolic fitness. AMPK-driven protection was associated with increased Sirt 3 expression and restoration of metabolic fitness. Renal AMPK activation in response to sepsis/IM is an adaptive mechanism that protects TEC, organs, and the host by preserving mitochondrial function and metabolic fitness likely through Sirt3 signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Carbonylcyanid-4-(trifluormethoxy)phenylhydrazon, ≥98% (HPLC), powder
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Supelco
Metformin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
ApopTag Peroxidase In-situ-Apoptosenachweis-Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
Tetramethylrhodamin-methylester-perchlorat, ≥95%
Sigma-Aldrich
Octoclothepin maleate salt, solid