Merck
  • The biophysical, molecular, and anatomical landscape of pigeon CRY4: A candidate light-based quantal magnetosensor.
The biophysical, molecular, and anatomical landscape of pigeon CRY4: A candidate light-based quantal magnetosensor.
Science advances (2020-08-28)
Tobias Hochstoeger, Tarek Al Said, Dante Maestre, Florian Walter, Alexandra Vilceanu, Miriam Pedron, Thomas D Cushion, William Snider, Simon Nimpf, Gregory Charles Nordmann, Lukas Landler, Nathaniel Edelman, Lennard Kruppa, Gerhard Dürnberger, Karl Mechtler, Stefan Schuechner, Egon Ogris, E Pascal Malkemper, Stefan Weber, Erik Schleicher, David A Keays
ZUSAMMENFASSUNG
The biophysical and molecular mechanisms that enable animals to detect magnetic fields are unknown. It has been proposed that birds have a light-dependent magnetic compass that relies on the formation of radical pairs within cryptochrome molecules. Using spectroscopic methods, we show that pigeon cryptochrome clCRY4 is photoreduced efficiently and forms long-lived spin-correlated radical pairs via a tetrad of tryptophan residues. We report that clCRY4 is broadly and stably expressed within the retina but enriched at synapses in the outer plexiform layer in a repetitive manner. A proteomic survey for retinal-specific clCRY4 interactors identified molecules that are involved in receptor signaling, including glutamate receptor-interacting protein 2, which colocalizes with clCRY4. Our data support a model whereby clCRY4 acts as an ultraviolet-blue photoreceptor and/or a light-dependent magnetosensor by modulating glutamatergic synapses between horizontal cells and cones.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Fetales Kälberserum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Millipore
Anti-FLAG® M2 Magnetperlen, affinity isolated antibody
Sigma-Aldrich
MEM Nicht-essentielle Aminosäure-Lösung (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Mayers Hämatoxylin-Lösung
Sigma-Aldrich
Chicken Serum, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
2-Phenylindol, technical grade, 95%