Merck
  • Sensitive colorimetric assay of carbidopa and methyldopa using tetrazolium blue chloride in pharmaceutical products.
Sensitive colorimetric assay of carbidopa and methyldopa using tetrazolium blue chloride in pharmaceutical products.
International Journal of Clinical Pharmacy (1989)
Issopoulos, P.B.