Merck
  • Total phytochemical analysis of Thymus munbyanus subsp. coloratus from Algeria by HS-SPME-GC-MS, NMR and HPLC-MSn studies.
Total phytochemical analysis of Thymus munbyanus subsp. coloratus from Algeria by HS-SPME-GC-MS, NMR and HPLC-MSn studies.
Journal of pharmaceutical and biomedical analysis (2020-05-07)
Hamdi Bendif, Gregorio Peron, Mohamed Djamel Miara, Stefania Sut, Stefano Dall'Acqua, Guido Flamini, Filippo Maggi
ZUSAMMENFASSUNG
In this article, we report a comprehensive characterization of volatile and polar constituents extracted from the aerial parts of Thymus munbyanus subsp. coloratus, a shrub that is used as culinary ingredient and as traditional medicine in Algeria, mainly to treat respiratory and gastrointestinal disorders and endocrine dysfunctions. Headspace solid phase microextraction (HS-SPME) coupled with gas chromatography-mass spectrometry (GC-MS) was used to assess volatile constituents, whereas the phytochemical composition of solid residues obtained from extraction with solvents at diffrent polarity was obtained by an integrated Nuclear Magnetic Resonance (NMR) and liquid chromatography coupled with tandem mass spectrometry (LC-MSn) approach. Fourty-five apolar ccompounds were identified, mainly oxygenated monoterpenes (65.8%), sesquiterpene hydrocarbons and nonoterpene hydrocarbons (18.6 and 14.5%, respectively). On the other hand, LC-MSn and NMR analyses revealed the presence of aglyconic and glycosilated flavonoids, phenylpropanoid derivatives and triterpenoid acids related to oleanolic acid, mainly in the methanol, dichloromethane and hexane extracts. Overall, these data indicate that Thymus munbyanus subsp. coloratus could be a potential source of antioxidants and bioactive compounds, and our results represent a starting point for further research on this plant species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)