Merck
Interstitial Cell Remodeling Promotes Aberrant Adipogenesis in Dystrophic Muscles.
Cell reports (2020-05-07)
Jordi Camps, Natacha Breuls, Alejandro Sifrim, Nefele Giarratana, Marlies Corvelyn, Laura Danti, Hanne Grosemans, Sebastiaan Vanuytven, Irina Thiry, Marzia Belicchi, Mirella Meregalli, Khrystyna Platko, Melissa E MacDonald, Richard C Austin, Rik Gijsbers, Giulio Cossu, Yvan Torrente, Thierry Voet, Maurilio Sampaolesi
ZUSAMMENFASSUNG
Fibrosis and fat replacement in skeletal muscle are major complications that lead to a loss of mobility in chronic muscle disorders, such as muscular dystrophy. However, the in vivo properties of adipogenic stem and precursor cells remain unclear, mainly due to the high cell heterogeneity in skeletal muscles. Here, we use single-cell RNA sequencing to decomplexify interstitial cell populations in healthy and dystrophic skeletal muscles. We identify an interstitial CD142-positive cell population in mice and humans that is responsible for the inhibition of adipogenesis through GDF10 secretion. Furthermore, we show that the interstitial cell composition is completely altered in muscular dystrophy, with a near absence of CD142-positive cells. The identification of these adipo-regulatory cells in the skeletal muscle aids our understanding of the aberrant fat deposition in muscular dystrophy, paving the way for treatments that could counteract degeneration in patients with muscular dystrophy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
bisBenzimid H 33342, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF488A F(ab′)2 fragment of antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Perilipin A/B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human GDF10