Merck
Comparison of IL-33 and IL-5 family mediated activation of human eosinophils.
PloS one (2019-09-07)
Evelyn L Angulo, Elizabeth M McKernan, Paul S Fichtinger, Sameer K Mathur
ZUSAMMENFASSUNG
Eosinophils are the prominent inflammatory cell involved in allergic asthma, atopic dermatitis, eosinophilic esophagitis, and hypereosinophilic syndrome and are found in high numbers in local tissue and/or circulating blood of affected patients. There is recent interest in a family of alarmins, including TSLP, IL-25 and IL-33, that are epithelial-derived and released upon stimulation of epithelial cells. Several genome wide association studies have found SNPs in genes encoding IL-33 to be risk factors for asthma. In two studies examining the direct role of IL-33 in eosinophils, there were differences in eosinophil responses. We sought to further characterize activation of eosinophils with IL-33 compared to activation by other cytokines and chemokines. We assessed IL-33 stimulated adhesion, degranulation, chemotaxis and cell surface protein expression in comparison to IL-3, IL-5, and eotaxin-1 on human eosinophils. Our results demonstrate that IL-33 can produce as potent eosinophil activation as IL-3, IL-5 and eotaxin-1. Thus, when considering specific cytokine targeting strategies, IL-33 will be important to consider for modulating eosinophil function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Chlorsuccinimid, 98%
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone N29, clone N29, Chemicon®, from mouse