Merck
Zebrafish miR-462-731 is required for digestive organ development.
Comparative biochemistry and physiology. Part D, Genomics & proteomics (2020-03-23)
Yan Huang, Chun-Xiao Huang, Wei-Feng Wang, Hong Liu, Huan-Ling Wang
ZUSAMMENFASSUNG
MicroRNAs (miRNAs), as important regulators of post-transcriptional gene expression, play important roles in the occurrence and function of organs. In this study, morpholino (MO) knockdown of miR-462/miR-731 was used to investigate the potential mechanisms of the miR-462-731 cluster during zebrafish liver development. The results showed significant reduction of digestive organs, especially liver and exocrine pancreas after the miR-462/miR-731 knockdown, and those phenotypes could be partially rescued by corresponding miRNA duplex. Acinar cells of the exocrine pancreas were also severely affected with pancreatic insufficiency. In particular, knockdown of miR-462 caused pancreas morphogenesis abnormity with specific bilateral exocrine pancreas. Additionally, it was found that miR-731 played a role in liver and exocrine pancreas development by directly targeting dkk3b, instead of the down-regulation of Wnt/β-catenin signaling. These findings contribute significantly to our understanding of molecular mechanisms of miR-462-731 cluster in development of digestive organs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
XAV939, ≥98% (HPLC)