Merck
Astrocyte glutamate transporters are increased in an early sporadic model of synucleinopathy.
Neurochemistry international (2020-05-23)
Luan Pereira Diniz, Ana Paula Bérgamo Araujo, Isadora Matias, Matheus N Garcia, Fernanda G Q Barros-Aragão, Ricardo Augusto de Melo Reis, Débora Foguel, Carolina Braga, Cláudia P Figueiredo, Luciana Romão, Flávia Carvalho Alcantara Gomes
ZUSAMMENFASSUNG
α-Synuclein protein (α-syn) is a central player in Parkinson's disease (PD) and in a spectrum of neurodegenerative diseases collectively known as synucleinopathies. These diseases are characterized by abnormal motor symptoms, such as tremor at rest, slowness of movement, rigidity of posture, and bradykinesia. Histopathological features of PD include preferential loss of dopaminergic neurons in the substantia nigra and formation of fibrillar intraneuronal inclusions called Lewy bodies and Lewy neurites, which are composed primarily of the α-syn protein. Currently, it is well accepted that α-syn oligomers (αSO) are the main toxic agent responsible for the etiology of PD. Glutamatergic excitotoxicity is associated with several neurological disorders, including PD. Excess glutamate in the synaptic cleft can be taken up by the astrocytic glutamate transporters GLAST and GLT-1. Although this event is the main defense against glutamatergic excitotoxicity, the molecular mechanisms that regulate this process have not yet been investigated in an early sporadic model of synucleinopathy. Here, using an early sporadic model of synucleinopathy, we demonstrated that the treatment of astrocytes with αSO increased glutamate uptake. This was associated with higher levels of GLAST and GLT-1 in astrocyte cultures and in a mouse model of synucleinopathy 24 h and 45 days after inoculation with αSO, respectively. Pharmacological inhibition of the TGF-β1 (transforming growth factor beta 1) pathway in vivo reverted GLAST/GLT-1 enhancement induced by αSO injection. Therefore, our study describes a new neuroprotective role of astrocytes in an early sporadic model of synucleinopathy and sheds light on the mechanisms of glutamate transporter regulation for neuroprotection against glutamatergic excitotoxicity in synucleinopathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade