Merck
  • The intestinal injury caused by ischemia-reperfusion is attenuated by amniotic fluid stem cells via the release of tumor necrosis factor-stimulated gene 6 protein.
The intestinal injury caused by ischemia-reperfusion is attenuated by amniotic fluid stem cells via the release of tumor necrosis factor-stimulated gene 6 protein.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-03-31)
Yuhki Koike, Bo Li, Carol Lee, Mashriq Alganabi, Haitao Zhu, Sinobol Chusilp, Dorothy Lee, Shigang Cheng, Qi Li, Agostino Pierro
ZUSAMMENFASSUNG
Ischemia/reperfusion (I/R) is implicated in the pathogenesis of various acute intestinal injuries. Amniotic fluid stem cells (AFSC) are beneficial in experimental intestinal diseases. Tumor necrosis factor-induced protein 6 (TSG-6) has been shown to exert anti-inflammatory effects. We aimed to investigate if AFSC secreted TSG-6 reduces inflammation and rescues intestinal I/R injury. The superior mesenteric artery of 3-week-old rats was occluded for 90 minutes and green fluorescent protein-labeled AFSC or recombinant TSG-6 was injected intravenously upon reperfusion. AFSC distribution was evaluated at 24, 48, and 72 hours after I/R. AFSC and TSG-6 effects on the intestine were assessed 48 hours postsurgery. Intestinal organoids were used to study the effects of TSG-6 after hypoxia-induced epithelial damage. After I/R-induced intestinal injury, AFSC migrated preferentially to the ileum, the primary site of injury, through blood circulation. Engrafted AFSC reduced ileum injury, inflammation, and oxidative stress. These AFSC-mediated beneficial effects were dependent on secretion of TSG-6. Administration of TSG-6 protected against hypoxia-induced epithelial damage in intestinal organoids. Finally, TSG-6 attenuated intestinal damage during I/R by suppressing genes involved in wound and injury pathways. This study indicates that AFSC or TSG-6 have the potential of rescuing the intestine from the damage caused by I/R.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFAIP6