Merck
  • Ubiquitin-Specific Protease 34 Inhibits Osteoclast Differentiation by Regulating NF-κB Signaling.
Ubiquitin-Specific Protease 34 Inhibits Osteoclast Differentiation by Regulating NF-κB Signaling.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2020-03-27)
Qiwen Li, Mengyuan Wang, Hanxiao Xue, Weiqing Liu, Yuchen Guo, Ruoshi Xu, Bin Shao, Quan Yuan
ZUSAMMENFASSUNG
The ubiquitination and deubiquitination enzymes ensure the stability and proper function of most cellular proteins. Disturbance of either enzyme compromises tissue homeostasis. We recently have identified that the ubiquitin-specific protease 34 (USP34) contributes to bone formation by promoting osteogenic differentiation of mesenchymal stem cells. However, its role in bone resorption, which couples bone formation, remains unknown. Here we show that knockdown of Usp34 promotes osteoclast differentiation of RAW264.7 cells. Conditional knockout of Usp34 in bone marrow-derived macrophages (BMMs) or in osteoclasts leads to elevated osteoclast function and low bone mass. Mechanically, we identify that USP34 restrains NF-κB signaling by deubiquitinating and stabilizing the NF-κB inhibitor alpha (IκBα). Overexpression of IκBα represses osteoclastic hyperfunction of Usp34-deficient RAW264.7 cells. Collectively, our results show that USP34 inhibits osteoclastogenesis by regulating NF-κB signaling. © 2020 American Society for Bone and Mineral Research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human USP34