Merck
  • Carbazole-Based Spiro[fluorene-9,9'-xanthene] as an Efficient Hole-Transporting Material for Perovskite Solar Cells.
Carbazole-Based Spiro[fluorene-9,9'-xanthene] as an Efficient Hole-Transporting Material for Perovskite Solar Cells.
ACS applied materials & interfaces (2020-06-02)
Do Yoon Lee, Gangala Sivakumar, Manju, Rajneesh Misra, Sang Il Seok
ZUSAMMENFASSUNG
For the practical application of perovskite solar cells (PSC), it is desirable to have high efficiency, long-term stability, and low manufacturing cost. Therefore, it is required to develop inexpensive and well-performing hole-transporting materials (HTMs). In this study, we synthesized SFXDAnCBZ, which is a new carbazole-based spiro[fluorene-9,9'-xanthene] (SFX) derivative, where the central core and end-cap units consist of SFX and N3,N6-bis(di-4-anisylamino)-9H-carbazole (DAnCBZ), respectively, as an efficient and low-cost HTM for PSCs. Photoluminescence quenching at the SFXDAnCBZ/perovskite interface was more effective than at the perovskite/Spiro-OMeTAD (2,2',7,7'-tetrakis-(N,N-di-p-methoxy-phenyl-amine) 9,9'spiro-bifluorene) interface. We fabricated a PSC with a power conversion efficiency (PCE) of 20.87% under 1 sun illumination (100 mW cm-2) using SFXDAnCBZ as an HTM. This value is comparable to that measured for the benchmark Spiro-OMeTAD. Thus, this result confirms that SFX core-based materials can be a new kind of HTMs for high-efficiency and low-cost PSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%