Merck
  • Milling interventions for the production of atta for Indian flat bread with low carbohydrate digestibility.
Milling interventions for the production of atta for Indian flat bread with low carbohydrate digestibility.
Journal of food science and technology (2020-07-31)
Tirupati Pawar, P Pavan Kumar, M S Ashwin Kumar, A Jyothi Lakshmi, Suresh D Sakhare, Indrani Dasappa, Aashitosh A Inamdar
ZUSAMMENFASSUNG
Study was aimed to produce atta for chapati, an Indian flat bread with low carbohydrate digestibility through different milling interventions; processing and formulating a functional ingredient mix (FM). Granulation, physico-chemical, rheological and chapati making characteristics of chakki atta, CA (control), roller mill atta (RA); RA replaced with 5, 10 and 15% FM (5, 10 and 15% RAFM) were evaluated. RA and RAFM samples showed lower water absorption, higher dough stability, pasting temperature and peak viscosity than CA. Evaluation of carbohydrate digestive profile showed differences in the pattern of carbohydrate digestibility and glucose release between the chapatis prepared from CA, RA and 10% RAFM. Rapidly available glucose (RAG), an indicator of glycemic response in vivo, was found to be lower in the 10% RAFM than CA. It can be concluded that milling interventions and compositional differences together determine the carbohydrate digestibility of the atta.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, ≥250 units/mg solid
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 8 × USP specifications
Sigma-Aldrich
Glucose-Oxidase aus Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Amyloglucosidase aus Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
Invertase aus Backhefe (S. cerevisiae), Grade VII, ≥300 units/mg solid
Sigma-Aldrich
Peroxidase aus Meerrettich, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
α-Amylase aus Bacillus amyloliquefaciens, liquid, ≥250 units/g