Merck
  • Combination of gel-electromembrane extraction with switchable hydrophilicity solvent-based homogeneous liquid-liquid microextraction followed by gas chromatography for the extraction and determination of antidepressants in human serum, breast milk and wastewater.
Combination of gel-electromembrane extraction with switchable hydrophilicity solvent-based homogeneous liquid-liquid microextraction followed by gas chromatography for the extraction and determination of antidepressants in human serum, breast milk and wastewater.
Journal of chromatography. A (2020-03-25)
Majid Behpour, Saeed Nojavan, Sakine Asadi, Ali Shokri
ZUSAMMENFASSUNG
The current study presents for the first time a combination of the gel electromembrane extraction (GEL-EME) and switchable hydrophilicity solvent-based homogeneous liquid-liquid microextraction (SHS-HLLME) methods which can be used as an efficient hyphenated extraction procedure. This coupled method, which was followed by GC-FID, was applied for quantification of antidepressants (desipramine, clozapine, and citalopram) in biological and wastewater samples. The effective parameters of both GEL-EME and SHS-HLLME procedures were optimized. Using an agarose gel membrane, analytes were extracted from 7.0 mL of the sample solution to 500 µL of the aqueous acceptor solution. The maximum extraction of analytes of interest was obtained under the optimized conditions (pH of acceptor solution, 5.0; pH of gel membrane, 5.0; pH of sample solution, 7.0, voltage value, 30 V; and extraction time, 30 min). Then, the acceptor solution was transferred to the extraction cell and the SHS-HLLME procedure was conducted again under the optimized conditions. Dipropylamine (50 µL) was selected as an extraction solvent. The introduced technique exhibited good linearities with coefficients of determinatin (R2) higher than 0.983 and an acceptable linear range of 5.0-1000 ng/mL. Accordingly, the limit of detection was ≤ 1.0 ng/mL (S/N = 3) for all analytes, and the high enrichment factors were obtained in the range of 178.7-194.8. Moreover, the corresponding repeatability was from 4.0 to 8.7% (n = 3). The proposed method was successfully utilized to determine trace levels of the drugs in human serum, wastewater, and breast milk samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dipropylamin, 99%
Sigma-Aldrich
2-(Diisopropylamino)ethylmethacrylat, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)