Merck
  • Frequency of MET exon 14 skipping mutations in non-small cell lung cancer according to technical approach in routine diagnosis: results from a real-life cohort of 2,369 patients.
Frequency of MET exon 14 skipping mutations in non-small cell lung cancer according to technical approach in routine diagnosis: results from a real-life cohort of 2,369 patients.
Journal of thoracic disease (2020-07-10)
Anne Champagnac, Pierre-Paul Bringuier, Marc Barritault, Sylvie Isaac, Emmanuel Watkin, Fabien Forest, Jean-Michel Maury, Nicolas Girard, Marie Brevet
ZUSAMMENFASSUNG
Mesenchymal epithelial transition receptor (MET) alterations, including MET exon 14 skipping mutation, are oncogenic in non-small cell lung cancer (NSCLC) and may confer sensitivity to targeted therapy. Given the rarity and the diversity of exon 14 skipping mutations, diagnosis may be challenging on small-biopsy specimens. Between March 2014 and May 2018, tissue samples from patients with metastatic NSCLC were analysed for MET exon 14 skipping mutation as part of routine practice in the Pathology Department of the Hospices Civils de Lyon, France. Over the study period, Sanger sequencing and/or two different DNA-based next generation sequencing (NGS) assays were used. Genomic alterations of MET exon 14 were detected in 2.6% (62/2,369) samples of NSCLC analysed for MET exon 14 mutations. Patients were mainly women (38/62, 61%) without smoking history (22/39, 56%) and the median age was 75 years. MET exon 14 skipping mutations were diagnosed by NGS in 50 cases and by classical Sanger sequencing in 12 cases. The frequency of MET mutations was 15.4% when Sanger sequencing was performed at the request of the clinician and 4.1% when the DNA-based NGS assay coverage included the 3' and 5' parts of the MET exon 14 and performed systematically. The frequency of genomic alterations is highly dependent on patient selection and the technical approach.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%