Merck
  • Growth performance, haematological responses, intestinal microbiology and carcass traits of broiler chickens fed finisher diets containing two-stage fermented banana peel meal.
Growth performance, haematological responses, intestinal microbiology and carcass traits of broiler chickens fed finisher diets containing two-stage fermented banana peel meal.
Tropical animal health and production (2019-11-25)
Sugiharto Sugiharto, Turrini Yudiarti, Isroli Isroli, Endang Widiastuti, Hanny Indrat Wahyuni, Tri A Sartono
ZUSAMMENFASSUNG
The study investigated the effect of finisher diets containing two-stage fermented banana peel (FBP) meal on growth performance, haematological responses, intestinal microbiology and carcass traits of broilers. From days 22 to 38, 200 broiler chicks were grouped to maize-soybean meal-based finisher diet as a control (CONT) and finisher diets containing either 5% FBP (FBP-5), 10% FBP (FBP-10) or 15% FBP (FBP-15). Birds were blood sampled and slaughtered at day 38. For microbial population and villi height measurement, digesta and intestinal segments were collected. Breast meat was obtained for meat colour determination. Data collected were analysed according to analysis of variance followed by Duncan's multiple-range test when there was significant effect. There was no effect of treatments on broiler growth performance. The relative duodenum weight tended (P = 0.08) to be lower in FBP than in CONT. The mean corpuscular haemoglobin concentration (MCHC) was higher (P < 0.05) in FBP-5 than in CONT and FBP-15. Uric acid concentration was lower (P < 0.05) in FBP-10 and FBP-15 than in CONT. Feeding FBP tended (P = 0.08) to decrease coliform population in the ileum. Compared to CONT, the ileal villi heights were higher (P < 0.05) and lower (P < 0.05) in FBP-10 and FBP-5, respectively. The relative weight of thigh (P < 0.05) and wings (P = 0.07) were higher in CONT than in FBP-fed birds, but the eviscerated carcass was not different (P > 0.05) among groups. The lightness (L*) values of meat were higher (P < 0.05) in FBP-10 and FBP-15 than in CONT and FBP-5. Compared with others, breast meat from FBP-10 birds had lower (P < 0.05) redness (a*) values. In conclusion, dietary inclusion of FBP at the levels of up to 15% had no detrimental effect on growth and health performances of broiler chickens.