Merck
A Deregulated HOX Gene Axis Confers an Epigenetic Vulnerability in KRAS-Mutant Lung Cancers.
Cancer cell (2020-04-04)
Stephanie L Guerra, Ophélia Maertens, Ryan Kuzmickas, Thomas De Raedt, Richard O Adeyemi, Caroline J Guild, Shawna Guillemette, Amanda J Redig, Emily S Chambers, Man Xu, Hong Tiv, Sandro Santagata, Pasi A Jänne, Stephen J Elledge, Karen Cichowski
ZUSAMMENFASSUNG
While KRAS mutations are common in non-small cell lung cancer (NSCLC), effective treatments are lacking. Here, we report that half of KRAS-mutant NSCLCs aberrantly express the homeobox protein HOXC10, largely due to unappreciated defects in PRC2, which confers sensitivity to combined BET/MEK inhibitors in xenograft and PDX models. Efficacy of the combination is dependent on suppression of HOXC10 by BET inhibitors. We further show that HOXC10 regulates the expression of pre-replication complex (pre-RC) proteins in sensitive tumors. Accordingly, BET/MEK inhibitors suppress pre-RC proteins in cycling cells, triggering stalled replication, DNA damage, and death. These studies reveal a promising therapeutic strategy for KRAS-mutant NSCLCs, identify a predictive biomarker of response, and define a subset of NSCLCs with a targetable epigenetic vulnerability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoklonaler Anti-β-Tubulin-Antikörper in Maus hergestellte Antikörper, clone TUB 2.1, ascites fluid