Merck
  • Interferon Alpha Favors Macrophage Infection by Visceral Leishmania Species Through Upregulation of Sialoadhesin Expression.
Interferon Alpha Favors Macrophage Infection by Visceral Leishmania Species Through Upregulation of Sialoadhesin Expression.
Frontiers in immunology (2020-06-26)
Lieselotte Van Bockstal, Dimitri Bulté, Magali Van den Kerkhof, Laura Dirkx, Dorien Mabille, Sarah Hendrickx, Peter Delputte, Louis Maes, Guy Caljon
ZUSAMMENFASSUNG
Type I interferons (IFNs) induced by an endogenous Leishmania RNA virus or exogenous viral infections have been shown to exacerbate infections with New World Cutaneous Leishmania parasites, however, the impact of type I IFNs in visceral Leishmania infections and implicated mechanisms remain to be unraveled. This study assessed the impact of type I IFN on macrophage infection with L. infantum and L. donovani and the implication of sialoadhesin (Siglec-1/CD169, Sn) as an IFN-inducible surface receptor. Stimulation of bone marrow-derived macrophages with type I IFN (IFN-α) significantly enhanced susceptibility to infection of reference laboratory strains and a set of recent clinical isolates. IFN-α particularly enhanced promastigote uptake. Enhanced macrophage susceptibility was linked to upregulated Sn surface expression as a major contributing factor to the infection exacerbating effect of IFN-α. Stimulation experiments in Sn-deficient macrophages, macrophage pretreatment with a monoclonal anti-Sn antibody or a novel bivalent anti-Sn nanobody and blocking of parasites with soluble Sn restored normal susceptibility levels. Infection of Sn-deficient mice with bioluminescent L. infantum promastigotes revealed a moderate, strain-dependent role for Sn during visceral infection under the used experimental conditions. These data indicate that IFN-responsive Sn expression can enhance the susceptibility of macrophages to infection with visceral Leishmania promastigotes and that targeting of Sn may have some protective effects in early infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polyinosinsäure:Polycytidylsäure Natriumsalz, TLR ligand tested