Merck
Chromatographic separation and automated analysis of flavanols.
Analytical biochemistry (1978-11-01)
I McMurrough, J McDowell
ZUSAMMENFASSUNG
Flavanols from barley or hops were separated chromatographically and assayed automatically by reaction with the chromogen, 4-dimethylaminocinnamaldehyde. For separating the flavanols on Sephadex G-25, gradient elution with water-methanol mixtures was necessary. The chromogen was specific for flavanols and well suited to AutoAnalyzer methods. The method appears generally applicable in flavanol analysis of plant materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(Dimethylamino)zimtaldehyd, chromogenic reagent for indoles and flavanols