Merck
  • Effects of drying processes on composition, microstructure and health aspects from maqui berries.
Effects of drying processes on composition, microstructure and health aspects from maqui berries.
Journal of food science and technology (2020-05-21)
Issis Quispe-Fuentes, Antonio Vega-Gálvez, Mario Aranda, Jacqueline Poblete, Alexis Pasten, Cristina Bilbao-Sainz, Delilah Wood, Tara McHugh, Carla Delporte
ZUSAMMENFASSUNG
The aim of this study is to determine the effects of different drying methods, including freeze drying (FD), convective drying, sun drying, infrared drying and vacuum drying (VD), on the chemical composition and microstructure of maqui berries as well as their anti-inflammatory and antidiabetic activities. Results showed that all dried samples have high unsaturated fatty acids contents (up to 83%) and high total dietary fiber contents (above 50%). Also, one hundred grams of dried berries provide between 11 and 21% of the recommended daily intake of α-tocopherol. Moreover, all dried maqui extracts reduced topical inflammation in treated mice. The highest anti-inflammatory effect against phorbol 12-myristate 13-acetate was found for VD and FD samples. Also, all dried maqui extracts showed antidiabetic activity by inhibiting α-glucosidase activity. The highest α-glucosidase inhibition activity was found for FD samples. The different biological activities of the dried maqui berries were related to differences in the extractability of metabolites due to microstructural changes during drying. The results indicated the potential use of dried maqui as a food ingredient with high unsaturated fatty acids, dietary fiber and α-tocopherol or as a natural extract with therapeutic agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Gesamtballaststoff-Assay-Kit, sufficient for ~100 assays
Sigma-Aldrich
α-Glucosidase aus Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Sigma-Aldrich
4-Nitrophenyl α-D-Glucopyranosid, ≥99%