Merck
  • How does substrate hydrophobicity affect the morphological features of reconstituted wax films and their interactions with nonionic surfactant and pesticide?
How does substrate hydrophobicity affect the morphological features of reconstituted wax films and their interactions with nonionic surfactant and pesticide?
Journal of colloid and interface science (2020-05-04)
Xuzhi Hu, Elias Pambou, Haoning Gong, Mingrui Liao, Peter Hollowell, Huayang Liu, Weimiao Wang, Carlo Bawn, Jos Cooper, Mario Campana, Kun Ma, Peixun Li, John R P Webster, Faheem Padia, Gordon Bell, Jian R Lu
ZUSAMMENFASSUNG
Surfactants are widely used in agri-sprays to improve pesticide efficiency, but the mechanism underlying their interactions with the surface wax film on plants remains poorly understood. To facilitate physical characterisations, we have reconstituted wheat cuticular wax films onto an optically flat silicon substrate with and without octadecyltrimethoxysilane modification to control surface hydrophobicity. Imaging techniques including scanning electron microscopy (SEM) unravelled morphological features of the reconstituted wax films similar to those on leaves, showing little impact from the different substrates used. Neutron reflection (NR) established that reconstituted wax films were comprised of an underlying wax film decorated with top surface wax protrusions, a common feature irrespective of substrate hydrophobicity and highly consistent with what was observed from natural wax films. NR measurements, with the help of isotopic H/D substitutions to modify the scattering contributions of the wax and solvent, revealed different wax regimes within the wax films, illustrating the impact of surface hydrophilicity on the nanostructures within the wax films. It was observed from both spectroscopic ellipsometry and NR measurements that wax films formed on the hydrophobic substrate were more robust and durable against attack by nonionic surfactant C12E6 solubilised with pesticide Cyprodinil (CP) than films coated on the bare hydrophilic silica. Thus, the former could be a more feasible model for studying the wax-surfactant-pesticide interactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimethoxy(octadecyl)silan, technical grade