Merck
  • Semiquantitative analysis of power doppler ultrasonography versus Tc-99m DMSA scintigraphy in diagnostic and severity assessment of acute childhood pyelonephritis.
Semiquantitative analysis of power doppler ultrasonography versus Tc-99m DMSA scintigraphy in diagnostic and severity assessment of acute childhood pyelonephritis.
Translational pediatrics (2020-09-22)
Hui Zhu, Minguang Chen, Hongxia Luo, Yin Pan, Wenjie Zheng, Yan Yang
ZUSAMMENFASSUNG
This study aimed to compare the diagnostic and predictive value of power Doppler ultrasonography (PDU) with Tc-99m dimercaptosuccinic acid (DMSA) renal scintigraphy in pediatric acute pyelonephritis (APN) using a semiquantitative analysis system. A total of 92 children and infants (184 kidneys) were hospitalized with possible APN. All children were examined by PDU and DMSA scintigraphy within 72 hours of admission. An empiric 9-point semiquantitative analysis system was used to sort kidneys into four grades (grade 0-III). Patients with several episodes of APN and congenital structural anomalies were excluded. Of 184 kidneys, we found 68 abnormal (grade I-III) and 116 normal (Grade 0) with DMSA scintigraphy, and 84 abnormal and 100 normal with PDU. In all, 23 kidneys were shown to be diseased by PDU but normal on DMSA scintigraphy while 7 kidneys showed the opposite trend. The sensitivity and specificity of PDU for diagnosing APN was 89.7% and 80.2%, respectively (P<0.05). In children older than 6 months, the sensitivity was higher (92%, P<0.05) than that in children younger than 6 months (87%, P<0.05). A moderate agreement (41%, P<0.05) on grade was found between the two methods. With the help of a semiquantitative analysis system, PDU can obviate the use of DMSA scintigraphy in children older than 6 months for APN diagnosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-2,3-Dimercaptobernsteinsäure, ~98%