Merck
  • CTDSP1 inhibitor rabeprazole regulates DNA-PKcs dependent topoisomerase I degradation and irinotecan drug resistance in colorectal cancer.
CTDSP1 inhibitor rabeprazole regulates DNA-PKcs dependent topoisomerase I degradation and irinotecan drug resistance in colorectal cancer.
PloS one (2020-08-09)
Hiroya Matsuoka, Koji Ando, Emma J Swayze, Elizabeth C Unan, Joseph Mathew, Quingjiang Hu, Yasuo Tsuda, Yuichiro Nakashima, Hiroshi Saeki, Eiji Oki, Ajit K Bharti, Masaki Mori
ZUSAMMENFASSUNG
Irinotecan specifically targets topoisomerase I (topoI), and is used to treat various solid tumors, but only 13-32% of patients respond to the therapy. Now, it is understood that the rapid rate of topoI degradation in response to irinotecan causes irinotecan resistance. We have published that the deregulated DNA-PKcs kinase cascade ensures rapid degradation of topoI and is at the core of the drug resistance mechanism of topoI inhibitors, including irinotecan. We also identified CTD small phosphatase 1 (CTDSP1) (a nuclear phosphatase) as a primary upstream regulator of DNA-PKcs in response to topoI inhibitors. Previous reports showed that rabeprazole, a proton pump inhibitor (PPI) inhibits CTDSP1 activity. The purpose of this study was to confirm the effects of rabeprazole on CTDSP1 activity and its impact on irinotecan-based therapy in colon cancer. Using differentially expressing CTDSP1 cells, we demonstrated that CTDSP1 contributes to the irinotecan sensitivity by preventing topoI degradation. Retrospective analysis of patients receiving irinotecan with or without rabeprazole has shown the effects of CTDSP1 on irinotecan response. These results indicate that CTDSP1 promotes sensitivity to irinotecan and rabeprazole prevents this effect, resulting in drug resistance. To ensure the best chance at effective treatment, rabeprazole may not be a suitable PPI for cancer patients treated with irinotecan.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human CTDSP1