Merck
  • Antidiabetic potential of gallic acid from Emblica officinalis: Improved glucose transporters and insulin sensitivity through PPAR-γ and Akt signaling.
Antidiabetic potential of gallic acid from Emblica officinalis: Improved glucose transporters and insulin sensitivity through PPAR-γ and Akt signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2019-05-09)
Bhavesh C Variya, Anita K Bakrania, Snehal S Patel
ZUSAMMENFASSUNG
Nature has gifted a variety of vital phytochemicals having potential therapeutic application against various ailments. Emblica officinalis (E. officinalis), an ancient plant, has long been used as a remedy for diabetes and cardiovascular complications, and presence of abundant amount of gallic acid could be accountable for its medicinal potential. The study was aimed to determine the in-vivo and in-vitro anti-diabetic potential of gallic acid and fruit juice of E. officinalis. Molecular mechanism of gallic acid as well as fruit juice of E. officinalis for anti-diabetic potential has also been revealed. Anti-diabetic potential of E. officinalis and gallic acid was evaluated in 3T3-L1 preadipocytes and various animal models like db/db mice and fructose administered rats. PPAR-γ expression and glucose translocation were observed using western blot and PCR techniques. Treatment of E. officinalis fruit juice and gallic acid facilitated their glucose homeostasis; improved insulin sensitivity; reduced obesity; abridged elevated blood pressure and declined cholesterol level, and also induced adipogenesis in 3T3-L1 adipocytes. Mechanistically, treatment increased expression of PPAR-γ through activation of C/EBPs and simultaneously increased Glut4 translocation in 3T3-L1 adipocytes. Moreover, gallic acid treatment increased insulin sensitivity through activation of Akt rather than AMPK signaling pathway while fruit juice of E. officinalis showed dual activation, Akt and AMPK as well. These findings reveal the role of gallic acid in E. officinalis mediated antidiabetic potential, and delineate the upregulation of pAkt, PPAR-γ and Glut4 in gallic acid mediated antidiabetic activity, thus providing potential therapy for diabetes and related disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
2-Desoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)