Merck
  • ATF4-mediated autophagy-dependent glycolysis plays an important role in attenuating apoptosis induced by Cr (VI) in A549 cells.
ATF4-mediated autophagy-dependent glycolysis plays an important role in attenuating apoptosis induced by Cr (VI) in A549 cells.
Toxicology letters (2020-06-23)
Zeyun Gao, Mongameli B Dlamini, Hong Ge, Liping Jiang, Chengyan Geng, Qiujuan Li, Xiaoxia Shi, Yong Liu, Jun Cao
ZUSAMMENFASSUNG
Chromium (Cr) (VI) compounds are known to be serious toxic and carcinogenic, but the mechanism is not clear. In our previous study, we found that Cr (VI)-induced ER stress plays an important role in the crosstalk between apoptosis and autophagy, while autophagy was apoptosis-dependent and subsequently prevents apoptosis cell death to keep A549 cells resistant to Cr (VI)-induced toxicity. In this study, we found that Cr (VI) could induce aerobic glycolysis in A549 cells. Both ER stress inhibitor, phenylbutyric acid (4-PBA) and the inhibitor of autophagy, 3-MA, repressed Cr (VI)-induced glycolysis, indicating that both ER stress and autophagy were involved in Cr (VI)-induced glycolysis in A549 cells. Co-treatment of the inhibitor of aerobic glycolysis, 2-DG and Cr (VI) for 24 h increased Cr (VI)-induced cleaved caspase-3, caspase-9 and the number of apoptotic cells, demonstrating that aerobic glycolysis played an important role in attenuating Cr (VI)-induced apoptosis. Furthermore, knockdown of ATF4 by siATF4 significantly decreased Cr (VI)-induced aerobic glycolysis and apoptosis, suggesting that ATF4 was involved in Cr (VI)-induced aerobic glycolysis and its effect of attenuating apoptosis in A549 cells. Taken together, our results demonstrated that autophagy-dependent glycolysis played a role in attenuating Cr (VI)-induced apoptosis. ER stress was involved in facilitating glycolysis, whose induction was mediated by ATF4. These findings open a window for the development of therapeutic interventions to prevent Cr (VI)-induced toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Immobilon®-P PVDF-Membran, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
bisBenzimid H 33342, suitable for fluorescence, ≥97.0% (HPLC)