Merck
  • SETD5-Coordinated Chromatin Reprogramming Regulates Adaptive Resistance to Targeted Pancreatic Cancer Therapy.
SETD5-Coordinated Chromatin Reprogramming Regulates Adaptive Resistance to Targeted Pancreatic Cancer Therapy.
Cancer cell (2020-05-23)
Zhentian Wang, Simone Hausmann, Ruitu Lyu, Tie-Mei Li, Shane M Lofgren, Natasha M Flores, Mary E Fuentes, Marcello Caporicci, Ze Yang, Matthew Joseph Meiners, Marcus Adrian Cheek, Sarah Ann Howard, Lichao Zhang, Joshua Eric Elias, Michael P Kim, Anirban Maitra, Huamin Wang, Michael Cory Bassik, Michael-Christopher Keogh, Julien Sage, Or Gozani, Pawel K Mazur
ZUSAMMENFASSUNG
Molecular mechanisms underlying adaptive targeted therapy resistance in pancreatic ductal adenocarcinoma (PDAC) are poorly understood. Here, we identify SETD5 as a major driver of PDAC resistance to MEK1/2 inhibition (MEKi). SETD5 is induced by MEKi resistance and its deletion restores refractory PDAC vulnerability to MEKi therapy in mouse models and patient-derived xenografts. SETD5 lacks histone methyltransferase activity but scaffolds a co-repressor complex, including HDAC3 and G9a. Gene silencing by the SETD5 complex regulates known drug resistance pathways to reprogram cellular responses to MEKi. Pharmacological co-targeting of MEK1/2, HDAC3, and G9a sustains PDAC tumor growth inhibition in vivo. Our work uncovers SETD5 as a key mediator of acquired MEKi therapy resistance in PDAC and suggests a context for advancing MEKi use in the clinic.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
meta-Phosphorsäure, ACS reagent, chips, 33.5-36.5%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Maisöl, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
3X FLAG® Peptid, lyophilized powder
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Millipore
Anti-V5 Agarose-Affinitätsgel in Maus hergestellte Antikörper, purified immunoglobulin, clone V5-10
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
Anti-Histon H4-Antikörper, pan, Klon 62-141-13, Kaninchen, monoklonal, clone 62-141-13, Upstate®, from rabbit
Sigma-Aldrich
UNC0638 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Protein-Arginin-N-Methyltransferasen-Inhibitor AMI-1, The Protein Arginine N-Methyltransferase Inhibitor, AMI-1 controls the biological activity of Protein Arginine N-Methyltransferase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)