Merck
  • lncRNA UCA1 Contributes to 5-Fluorouracil Resistance of Colorectal Cancer Cells Through miR-23b-3p/ZNF281 Axis.
lncRNA UCA1 Contributes to 5-Fluorouracil Resistance of Colorectal Cancer Cells Through miR-23b-3p/ZNF281 Axis.
OncoTargets and therapy (2020-08-18)
Zhenyu Xian, Bang Hu, Ting Wang, Junyi Zeng, Jinlin Cai, Qi Zou, Peixuan Zhu
ZUSAMMENFASSUNG
The chemoresistance of 5-fluorouracil (5-FU) limited the application of chemotherapy in colorectal cancer (CRC) treatment. Herein, we aimed to uncover the potential mechanism behind the 5-FU resistance of CRC cells. The abundance of long noncoding RNA urothelial carcinoma associated 1 (lncRNA UCA1), microRNA-23b-3p (miR-23b-3p) and zinc finger protein 281 (ZNF281) was measured by quantitative real-time polymerase chain reaction (qRT-PCR) in CRC tissues and cells. Western blot was conducted to examine autophagy-related proteins, apoptosis-associated proteins and ZNF281 in CRC tissues and cells. Cell counting kit-8 (CCK8) assay was performed to detect the viability and inhibitory concentration 50% (IC50) value of 5-FU of CRC cells. The apoptosis of CRC cells was measured by flow cytometry. The binding sites between miR-23b-3p and UCA1 or ZNF281 were predicted by miRcode and Starbase software, respectively, and the combination was confirmed by dual-luciferase reporter assay and RIP assay. Murine xenograft model was established to verify the role of UCA1 on the 5-FU resistance of CRC in vivo. The 5-FU resistance of CRC was positively related to the level of UCA1 and autophagy. UCA1 accelerated the 5-FU resistance of CRC cells through facilitating autophagy and suppressing apoptosis. MiR-23b-3p was a target of UCA1 in 293T and CRC cells. The knockdown of miR-23b-3p reversed the inhibitory effects of UCA1 interference on the 5-FU resistance and autophagy and the promoting impact on the apoptosis of CRC cells. ZNF281 could bind to miR-23b-3p in 293T cells. MiR-23b-3p elevated the 5-FU sensitivity through down-regulating ZNF281 in CRC cells. UCA1 interference enhanced the 5-FU sensitivity of CRC through miR-23b-3p/ZNF281 axis in vivo. UCA1 mediated 5-FU resistance of CRC cells through facilitating autophagy and inhibiting apoptosis via miR-23b-3p/ZNF281 axis in vivo and in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human ZNF281