Merck
A broad-spectrum antibiotic adjuvant reverses multidrug-resistant Gram-negative pathogens.
Nature microbiology (2020-05-20)
Meirong Song, Yuan Liu, Xiaoyong Huang, Shuangyang Ding, Yang Wang, Jianzhong Shen, Kui Zhu
ZUSAMMENFASSUNG
The rapid emergence and dissemination of multidrug-resistant (MDR) bacterial pathogens pose a serious threat to global healthcare. One particular concern is the carbapenem-resistant Enterobacteriaceae (CRE), a group of Gram-negative bacteria that have evolved resistance to all or nearly all available antibiotics. Coupled with the fact of barren antibiotic development pipeline nowadays, a critical approach is to revitalize existing antibiotics using antibiotic adjuvants. We found a short linear antibacterial peptide (SLAP)-S25 carrying four non-natural amino acids of 2,4-diaminobutanoic acid (Dab), which solely showed weak antibacterial activity but boosted the efficacy of antibiotics covering all major classes, including cefepime, colistin, ofloxacin, rifampicin, tetracycline and vancomycin, against MDR Gram-negative pathogens. Mechanistic studies showed that SLAP-S25 triggers membrane damage by binding to both lipopolysaccharide (LPS) in the outer membrane and phosphatidylglycerol (PG) in bacterial cytoplasmic membrane, to potentiate antibiotic efficacy through collaborative strategies. Lastly, SLAP-S25 effectively enhanced the activity of colistin against MDR Escherichia coli-associated infections in three animal models. Our findings provide a potential therapeutic option using existing antibiotics in combination with broad-spectrum antibiotic adjuvants, to address the prevalent infections caused by MDR Gram-negative pathogens worldwide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
N-Phenyl-1-naphthylamin, reagent grade, 98%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Octoclothepin maleate salt, solid