Merck
  • High-fat diet pre-conditioning improves microvascular remodelling during regeneration of ischaemic mouse skeletal muscle.
High-fat diet pre-conditioning improves microvascular remodelling during regeneration of ischaemic mouse skeletal muscle.
Acta physiologica (Oxford, England) (2020-02-06)
Emmanuel Nwadozi, Martina Rudnicki, Matthew De Ciantis, Stephanie Milkovich, Alexandru Pulbere, Emilie Roudier, Olivier Birot, Thomas Gustafsson, Christopher G Ellis, Tara L Haas
ZUSAMMENFASSUNG
Critical limb ischaemia (CLI) is characterized by inadequate angiogenesis, arteriolar remodelling and chronic myopathy, which are most severe in type 2 diabetic patients. Hypertriglyceridaemia, commonly observed in these patients, compromises macrovascular function. However, the effects of high-fat diet-induced increases in circulating lipids on microvascular remodelling are not established. Here, we investigated if high-fat diet would mimic the detrimental effect of type 2 diabetes on post-ischaemia vascular remodelling and muscle regeneration, using a mouse model of hindlimb ischaemia. Male C57Bl6/J mice were fed with normal or high-fat diets for 8 weeks prior to unilateral femoral artery ligation. Laser doppler imaging was used to assess limb perfusion recovery. Vascular recovery, inflammation, myofibre regeneration and fibrosis were assessed at 4 or 14 days post-ligation by histology and RNA analyses. Capillary-level haemodynamics were assessed by intravital microscopy of control and regenerating muscles 14 days post-ligation. High-fat diet increased muscle succinate dehydrogenase activity and capillary-level oxygen supply. At 4 days post-ligation, no diet differences were detected in muscle damage, inflammatory infiltration or capillary activation. At 14 days post-ligation, high fat-fed mice displayed accelerated limb blood flow recovery, elevated capillary and arteriole densities as well as greater red blood cell supply rates and capillary-level oxygen supply. Regenerating muscles from high fat-fed mice displayed lower interstitial fat and collagen deposition. The muscle-level adaptations to high-fat diet improved multiple aspects of muscle recovery in response to ischaemia and did not recapitulate the worse outcomes seen in diabetic CLI patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin, α-Glattmuskel - Cy3 Maus-Antikörper, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Direktrot 80, Dye content 25 %