Merck
  • Demethoxycurcumin analogue DMC-BH inhibits orthotopic growth of glioma stem cells by targeting JNK/ERK signaling.
Demethoxycurcumin analogue DMC-BH inhibits orthotopic growth of glioma stem cells by targeting JNK/ERK signaling.
Aging (2020-07-28)
Lei Shi, Guan Sun, Haifeng Zhu
ZUSAMMENFASSUNG
Glioma stem cells (GSCs) play an important role in glioblastoma resistance to conventional therapies and disease recurrence. Here, we assessed the therapeutic effect of a demethoxycurcumin analogue, DMC-BH, on GSCs, and investigated the underlying mechanisms. Our in vitro data demonstrate that DMC-BH inhibits GSC proliferation, and induces apoptosis and autophagy in GSCs. In addition, our results show that DMC-BH effectively crosses the blood-brain barrier to inhibit the growth of intracranial GSC tumors in vivo. DMC-BH significantly increased phosphorylation levels of JNK, ERK and c-Jun in GSCs. Inhibition of JNK and ERK activities reversed the pro-apoptotic effect of DMC-BH in GSCs, indicating that the DMC-BH-induced apoptosis in GSCs is mediated via the JNK/ERK signaling pathway. These results suggest that DMC-BH could potentially serve as a effective therapy against GSCs that acts by targeting the JNK/ERK signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, ≥97% (HPLC), powder