Merck
  • Newly discovered Synechococcus sp. PCC 11901 is a robust cyanobacterial strain for high biomass production.
Newly discovered Synechococcus sp. PCC 11901 is a robust cyanobacterial strain for high biomass production.
Communications biology (2020-05-10)
Artur Włodarczyk, Tiago Toscano Selão, Birgitta Norling, Peter J Nixon
ZUSAMMENFASSUNG
Cyanobacteria, which use solar energy to convert carbon dioxide into biomass, are potential solar biorefineries for the sustainable production of chemicals and biofuels. However, yields obtained with current strains are still uncompetitive compared to existing heterotrophic production systems. Here we report the discovery and characterization of a new cyanobacterial strain, Synechococcus sp. PCC 11901, with promising features for green biotechnology. It is naturally transformable, has a short doubling time of ≈2 hours, grows at high light intensities and in a wide range of salinities and accumulates up to ≈33 g dry cell weight per litre when cultured in a shake-flask system using a modified growth medium - 1.7 to 3 times more than other strains tested under similar conditions. As a proof of principle, PCC 11901 engineered to produce free fatty acids yielded over 6 mM (1.5 g L-1), an amount comparable to that achieved by similarly engineered heterotrophic organisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridiniumchlorchromat, 98%