Merck
  • Simultaneous Analysis of Fenthion and Its Five Metabolites in Produce Using Ultra-High Performance Liquid Chromatography-Tandem Mass Spectrometry.
Simultaneous Analysis of Fenthion and Its Five Metabolites in Produce Using Ultra-High Performance Liquid Chromatography-Tandem Mass Spectrometry.
Molecules (Basel, Switzerland) (2020-04-26)
Jonghwa Lee, Jeong-Han Kim
ZUSAMMENFASSUNG
A simultaneous analytical method for the organophosphorus insecticide fenthion and its five metabolites (fenthion oxon, fenthion oxon sulfoxide, fenthion oxon sulfone, fenthion sulfoxide, and fenthion sulfone) was developed based on ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). Five matrices (brown rice, chili pepper, orange, potato, and soybean) were selected to validate the method. The target compounds were analyzed using positive electrospray ionization in the multiple reaction monitoring mode. For the best sensitivity in regard to the detector response, water and methanol containing formic acid (0.1%) were selected as the mobile phase. The optimum extraction efficiency was obtained through a citrate-buffered QuEChERS (quick, easy, cheap, effective, rugged, and safe) method. Recovery tests were carried out at three spiking levels (n = 3). At all fortification levels, the accuracy and precision results were between 70% and 120% with a relative standard deviation of ≤15%. The limit of quantitation was 0.01 mg/kg, and the correlation coefficients (r2) of the matrix-matched calibration curves were >0.99. Significant signal suppression in the detector responses were observed for all matrices, suggesting that a compensation method, such as matrix-matched calibration, is required to provide accurate quantitative results. The applicability of the presented method was confirmed for the simultaneous analysis of fenthion and its metabolites in various crops.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Fenthion-sulfon, PESTANAL®, analytical standard