Merck
  • In vitro assessment of pesticide residues bioaccessibility in conventionally grown blueberries as affected by complex food matrix.
In vitro assessment of pesticide residues bioaccessibility in conventionally grown blueberries as affected by complex food matrix.
Chemosphere (2020-03-30)
Danijel D Milinčić, Uroš D Vojinović, Aleksandar Ž Kostić, Mirjana B Pešić, Bojana D Špirović Trifunović, Dragica V Brkić, Milan Ž Stević, Milan O Kojić, Nemanja S Stanisavljević
ZUSAMMENFASSUNG
The aim of this study was to investigate the bioaccessibility of pesticide residues in blueberries (commercial and sample from controlled field trial) from Serbia, involving the presence of a complex food matrix and to assess the potential risk to human health. The presence of nine active substances (azoxystrobin, boscalid, fludioxonil, cyprodinil, pyrimethanil, pyridaben, pyriproxyfen, acetamiprid and thiametoxam) in initial blueberry samples was determined in concentration range from 5.15 μg/kg for thiametoxam to 187 μg/kg for azoxystrobin. Clothianidin, metabolite of thiametoxam, was not detected in any blueberry sample. However, after in vitro digestion, the content of initially detected pesticides residues was significantly decreased or it was below limit of quantification resulting in the total bioaccessibility of about 15%. Azoxystrobin, pyrimethanil and fludioxonil was quantified in digestive juice at concentrations which were about 81%, 37% and 10% less than the inital concentration, respectively. The presence of food matrix during digestion of blueberries even more severely reduced concentration of pesticide residues (total bioaccessibility was about 7%) compared to digestion without the food matrix. Only azoxystrobin was quantified after digestion with food matrix in concentration of 27 μg/kg in sample from controlled field trial and detected in two commercial samples but below the limit of quantification. Furthermore, chronic risk assessment indicated that risk is acceptable for the health of different human subpopulation groups. The current study on pesticides residues, most commonly applied on blueberries, provides for the first time an insight into their bioaccessibility under conditions that mimic physiological environment of human digestive tract.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, ≥250 units/mg solid
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 4 × USP specifications
Sigma-Aldrich
Bile extract porcine
Supelco
Fludioxonil, PESTANAL®, analytical standard
Supelco
Azoxystrobin, PESTANAL®, analytical standard
Supelco
Pyriproxyfen, PESTANAL®, analytical standard
Supelco
Trifloxystrobin, PESTANAL®, analytical standard
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
Fluopyram, PESTANAL®, analytical standard
Supelco
N-(6-Chlor-3-pyridylmethyl)-N-cyan-N-methylacetamidin, PESTANAL®, analytical standard
Supelco
Cyantraniliprol, PESTANAL®, analytical standard
Supelco
Pyrimethanil, PESTANAL®, analytical standard
Supelco
Carbofuran-d3, PESTANAL®, analytical standard