Merck
  • Liquid Chromatography Mass Spectrometry Quantification of α-solanine, α-chaconine, and Solanidine in Potato Protein Isolates.
Liquid Chromatography Mass Spectrometry Quantification of α-solanine, α-chaconine, and Solanidine in Potato Protein Isolates.
Foods (Basel, Switzerland) (2020-04-08)
Søren D Nielsen, Jesper M Schmidt, Gitte H Kristiansen, Trine K Dalsgaard, Lotte B Larsen
ZUSAMMENFASSUNG
For potato proteins to be used as a food ingredient, the level of natural potato defense substances, the glycoalkaloids (GAs), should be limited. In this work, a method is developed for quantification of the two major potato GAs, α-solanine and α-chaconine, as well as for their aglycon form, solanidine, using liquid chromatography-mass spectrometry single quadrupole in single ion monitoring mode. Standard solutions of GA and a food-grade potato protein powder was used to validate the method. A linear correlation between GA concentration and the ion intensity of >0.995 was obtained for all standard solutions. Recovery of GA in spiked samples was within the range 82%-106%. The method for GA quantification was applied to a variety of potato protein isolates. The results showed that total GA increased during the storage of the potatoes. Washing the potato protein isolates using water at a sufficient level was shown to be able to reduce the amount of GA below the threshold of 150 µg g-1, as needed for human consumption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Solanidin, ≥97.0% (HPLC)