Merck
Galectin-3 modulates epithelial cell adaptation to stress at the ER-mitochondria interface.
Cell death & disease (2020-05-14)
Lucie Coppin, Arnaud Jannin, Emilie Ait Yahya, Caroline Thuillier, Céline Villenet, Meryem Tardivel, Antonino Bongiovanni, Cécile Gaston, Simon de Beco, Nicolas Barois, Isabelle van Seuningen, Emmanuelle Durand, Amélie Bonnefond, Jean-Claude Vienne, Joseph Vamecq, Martin Figeac, Audrey Vincent, Delphine Delacour, Nicole Porchet, Pascal Pigny
ZUSAMMENFASSUNG
Cellular stress response contributes to epithelial defense in adaptation to environment changes. Galectins play a pivotal role in the regulation of this response in malignant cells. However, precise underlying mechanisms are largely unknown. Here we demonstrate that Galectin-3, a pro and anti-apoptotic lectin, is required for setting up a correct cellular response to stress by orchestrating several effects. First, Galectin-3 constitutes a key post-transcriptional regulator of stress-related mRNA regulons coordinating the cell metabolism, the mTORC1 complex or the unfolded protein response (UPR). Moreover, we demonstrated the presence of Galectin-3 with mitochondria-associated membranes (MAM), and its interaction with proteins located at the ER or mitochondrial membranes. There Galectin-3 prevents the activation and recruitment at the mitochondria of the regulator of mitochondria fission DRP-1. Accordingly, loss of Galectin-3 impairs mitochondrial morphology, with more fragmented and round mitochondria, and dynamics both in normal and cancer epithelial cells in basal conditions. Importantly, Galectin-3 deficient cells also display changes of the activity of the mitochondrial respiratory chain complexes, of the mTORC1/S6RP/4EBP1 translation pathway and reactive oxygen species levels. Regarding the ER, Galectin-3 did not modify the activities of the 3 branches of the UPR in basal conditions. However, Galectin-3 favours an adaptative UPR following ER stress induction by Thapsigargin treatment. Altogether, at the ER-mitochondria interface, Galectin-3 coordinates the functioning of the ER and mitochondria, preserves the integrity of mitochondrial network and modulates the ER stress response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Assay-Kit zur Untersuchung des ADP/ATP-Verhältnisses, sufficient for 100 tests (bioluminescent)
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)