Merck
  • Prevalence of atrial fibrillation before cardiac surgery and factors associated with concomitant ablation.
Prevalence of atrial fibrillation before cardiac surgery and factors associated with concomitant ablation.
The Journal of thoracic and cardiovascular surgery (2019-08-25)
Patrick M McCarthy, Charles J Davidson, Jane Kruse, Daniel J Lerner, Mary Jo Braid-Forbes, Michelle M McCrea, Ahmed M Elmouelhi, Michael A Ferguson
ZUSAMMENFASSUNG
Surgical ablation for atrial fibrillation concomitant with cardiac surgery is now a Class I recommendation for selected patients. Understanding how the revised recommendations will affect appropriate use of surgical ablation is challenging because the reported prevalence of preoperative atrial fibrillation depends on the definition used. The objective was to determine the prevalence of atrial fibrillation in the 3 years before cardiac surgery and the rate of concomitant surgical ablation. Patients with and without a diagnosis of atrial fibrillation in the 3 years before surgical coronary artery bypass, aortic valve replacement, or mitral valve replacement/repair were identified in the 2014 Medicare Standard Analytical File. Patients had prior atrial fibrillation in 28.4% of 79,134 cardiac surgeries. Prior atrial fibrillation was associated with risk factors for increased cardiac surgical morbidity/mortality, including recent heart failure, renal failure, and lung disease. Black patients were less likely to have prior atrial fibrillation but more likely to have had infrequent care for it. Isolated coronary artery bypass had the lowest prevalence but highest absolute number of prior atrial fibrillation cases. Concomitant surgical ablation was performed in 22.1% of patients with prior atrial fibrillation. Patients with mitral valve surgery were 3-fold more likely to receive surgical ablation. Women were less likely to have prior atrial fibrillation but less likely to have surgical ablation when they did. Medicare beneficiaries had a substantially higher prevalence of atrial fibrillation diagnoses in the 3 years before cardiac surgery than previously published rates of preoperative atrial fibrillation. Concomitant surgical ablation was performed in less than one-quarter of patients with atrial fibrillation undergoing cardiac surgery for other indications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder