Merck
Glia-to-Neuron Conversion by CRISPR-CasRx Alleviates Symptoms of Neurological Disease in Mice.
Cell (2020-04-10)
Haibo Zhou, Jinlin Su, Xinde Hu, Changyang Zhou, He Li, Zhaorong Chen, Qingquan Xiao, Bo Wang, Wenyan Wu, Yidi Sun, Yingsi Zhou, Cheng Tang, Fei Liu, Linhan Wang, Canbin Feng, Mingzhe Liu, Sanlan Li, Yifeng Zhang, Huatai Xu, Haishan Yao, Linyu Shi, Hui Yang
ZUSAMMENFASSUNG
Conversion of glial cells into functional neurons represents a potential therapeutic approach for replenishing neuronal loss associated with neurodegenerative diseases and brain injury. Previous attempts in this area using expression of transcription factors were hindered by the low conversion efficiency and failure of generating desired neuronal types in vivo. Here, we report that downregulation of a single RNA-binding protein, polypyrimidine tract-binding protein 1 (Ptbp1), using in vivo viral delivery of a recently developed RNA-targeting CRISPR system CasRx, resulted in the conversion of Müller glia into retinal ganglion cells (RGCs) with a high efficiency, leading to the alleviation of disease symptoms associated with RGC loss. Furthermore, this approach also induced neurons with dopaminergic features in the striatum and alleviated motor defects in a Parkinson's disease mouse model. Thus, glia-to-neuron conversion by CasRx-mediated Ptbp1 knockdown represents a promising in vivo genetic approach for treating a variety of disorders due to neuronal loss.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Tyrosin-Hydroxylase-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
6-Hydroxydopamin -hydrochlorid, ≥97% (titration), powder
Sigma-Aldrich
Anti-NeuN-Antikörper (Kaninchen), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN-Antikörper, serum, from guinea pig
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Sigma-Aldrich
Anti-Dopamin-Transporter-Antikörper, NT, Klon DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®
Sigma-Aldrich
Desipramin -hydrochlorid, ≥98% (TLC), powder
Sigma-Aldrich
R-(−)-Apomorphin -hydrochlorid Hemihydrat, calcined, ≥98% (with NaOH, titration)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)