Merck
Carbon Dots@rGO Paper as Freestanding and Flexible Potassium-Ion Batteries Anode.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2020-08-11)
Erjin Zhang, Xinxin Jia, Bin Wang, Jue Wang, Xinzhi Yu, Bingan Lu
ZUSAMMENFASSUNG
Carbonaceous materials, especially with graphite-layers structure, as anode for potassium-ion batteries (PIBs), are the footstone for industrialization of PIBs. However, carbonaceous materials with graphite-layers structure usually suffer from poor cycle life and inferior stability, not to mention freestanding and flexible PIBs. Here, a freestanding and flexible 3D hybrid architecture by introducing carbon dots on the reduced graphene oxide surface (CDs@rGO) is synthesized as high performance PIBs anode. The CDs@rGO paper has efficient electron and ion transfer channels due to its unique structure, thus enhancing reaction kinetics. In addition, the CDs provide abundant defects and oxygen-containing functional groups, which can improve the electrochemical performance. This freestanding and flexible anode exhibits the high capacity of 310 mAh g-1 at 100 mA g-1, ultra-long cycle life (840 cycles with a capacity of 244 mAh g-1 at 200 mA g-1), and excellent rate performance (undergo six consecutive currents changing from 100 to 500 mA g-1, high capacity 185 mAh g-1 at 500 mA g-1), outperforming many existing carbonaceous PIB anodes. The results may provide a starting point for high-performance freestanding and flexible PIBs and promote the rapid development of next-generation flexible batteries.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Yttrium-Aluminium-Oxid, nanopowder, <150 nm particle size (TEM), 99% trace metals basis