Merck
  • Obtainment and Characterization of Hydrophilic Polysulfone Membranes by N-Vinylimidazole Grafting Induced by Gamma Irradiation.
Obtainment and Characterization of Hydrophilic Polysulfone Membranes by N-Vinylimidazole Grafting Induced by Gamma Irradiation.
Polymers (2020-06-10)
Elizabeth Vázquez, Claudia Muro, Javier Illescas, Guillermina Burillo, Omar Hernández, Ernesto Rivera
ZUSAMMENFASSUNG
Polysulfone (PSU) film and N-vinylimidazole (VIM) were used to obtain grafted membranes with high hydrophilic capacity. The grafting process was performed by gamma irradiation under two experiments: (1) different irradiation doses (100-400 kGy) and VIM 50% solution; (2) different concentration of grafted VIM (30-70%) and 300 kGy of irradiation dose. Characteristics of the grafted membranes were determined by Fourier transform infrared spectroscopy (FTIR), scanning electron microscopy (SEM), contact angle, swelling degree, desalination test, thermogravimetric analysis (TGA) and differential scanning calorimetry (DSC). Both experiments indicated that the absorbed dose 300 kGy and the VIM concentration, at 50% v/v, were effective to obtain PSU grafted membranes with 14.3% of grafting yield. Nevertheless, experimental conditions, 400 kGy, VIM 50% and 300 kGy, VIM 60-70% promoted possible membrane degradation and VIM homopolymerization on the membrane surface, which was observed by SEM images; meanwhile, 100-200 kGy and VIM 30-50% produced minimal grafting (2 ± 0.5%). Hydrophilic surface of the grafted PSU membranes by 300 kGy and VIM 50% v/v were corroborated by the water contact angle, swelling degree and desalination test, showing a decrease from 90.7° ± 0.3 (PSU film) to 64.3° ± 0.5; an increment of swelling degree of 25 ± 1%, and a rejection-permeation capacity of 75 ± 2%. In addition, the thermal behavior of grafted PSU membranes registered an increment in the degradation of 20%, due to the presence of VIM. However, the normal temperature of the membrane operation did not affect this result; meanwhile, the glass transition temperature (Tg) of the grafted PSU membrane was found at 185.4 ± 0.5 °C, which indicated an increment of 15 ± 1%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Vinylimidazol, ≥99%