Merck
Streptavidin-Hosted Organocatalytic Aldol Addition.
Molecules (Basel, Switzerland) (2020-05-30)
Nicolò Santi, Louis C Morrill, Louis Y P Luk
ZUSAMMENFASSUNG
In this report, the streptavidin-biotin technology was applied to enable organocatalytic aldol addition. By attaching pyrrolidine to the valeric motif of biotin and introducing it to streptavidin (Sav), a protein-based organocatalytic system was created, and the aldol addition of acetone with p-nitrobenzaldehyde was tested. The conversion of substrate to product can be as high as 93%. Although the observed enantioselectivity was only moderate (33:67 er), further protein engineering efforts can be included to improve the selectivity. These results have proven the concept that Sav can be used to host stereoselective aldol addition.