Merck
  • Leaching of brominated flame retardants (BFRs) from BFRs-incorporated plastics in digestive fluids and the influence of bird diets.
Leaching of brominated flame retardants (BFRs) from BFRs-incorporated plastics in digestive fluids and the influence of bird diets.
Journal of hazardous materials (2020-03-03)
Huiying Guo, Xiaobo Zheng, Xiaojun Luo, Bixian Mai
ZUSAMMENFASSUNG
Leaching kinetics of additive-derived brominated flame retardants (BFRs) in different sizes (100 μm-2 mm) of acrylonitrile-butadiene-styrene copolymer (ABS) plastics were investigated in water, simulated gastric fluids, and simulated gastrointestinal fluids. The influences of bird diets (fish, clam, and rice) on the leaching of BFRs from plastics were also explored. The leaching kinetics of BFRs were best fitted with the second-order diffusion model. The leaching rates of BFRs increased for the less lipophilic BFRs in finer sizes of ABS. The log-transformed leached proportions of BFRs at equilibrium were significantly correlated with logKOW of BFRs (p < 0.05). BFRs migrated from ABS to digestive fluids and diet residues at equilibrium, since elevated concentrations of BFRs were observed in diet residues than virgin diet samples. Leached proportions of BFRs in gut fluids from mixture of ABS and diets were lower than those from only ABS. The logKOW of BFRs and the migration proportions of BFRs from ABS to digestive fluids and diet residues were fitted with linear regression analysis. The results indicate that more lipophilic BFRs are preferentially leached from BFRs-incorporated plastics into fluids and are further adsorbed by diet residues.