Merck
  • Design, synthesis and biological evaluation of tetrahydroquinoline-based reversible LSD1 inhibitors.
Design, synthesis and biological evaluation of tetrahydroquinoline-based reversible LSD1 inhibitors.
European journal of medicinal chemistry (2020-04-02)
Xinran Wang, Cai Zhang, Xiangyu Zhang, Jiangkun Yan, Jiming Wang, Qinwen Jiang, Liyu Zhao, Dongmei Zhao, Maosheng Cheng
ZUSAMMENFASSUNG
The targeted regulation of LSD1, which is highly expressed in a variety of tumor cells, is a promising cancer therapy strategy. Several LSD1 inhibitors are currently under clinical evaluation, and most of these inhibitors are irreversible. Here, we report the design, synthesis and biochemical evaluation of novel tetrahydroquinoline-based reversible LSD1 inhibitors. Compounds 18s and 18x, which are selective to LSD1 over MAO-A/B, exhibit excellent LSD1 inhibition at the molecular levels with IC50 = 55 nM and 540 nM, respectively. The classic Lineweaver-Burk plots revealed that compound 18s could reversibly bind the LSD1 enzyme in a noncompetitive manner. Molecular docking was used to reveal the potential binding-mode of the compounds and interpret the structure-activity relationships. Furthermore, compounds 18s and 18x significantly inhibited proliferation (IC50 = 1.13 μM and 1.15 μM, respectively) and induced apoptosis in MGC-803 cells with high expression of LSD1. Compound 18x showed acceptable liver microsomal stability. Meanwhile, 18x did not appear to inhibit CYPs at 10 μM in vitro. Remarkably, the oral administration of compound 18x can inhibit the growth of MGC-803 xenograft tumors without significant side effects. Our findings suggest that tetrahydroquinoline-based LSD1 inhibitors deserve further investigation for the treatment of LSD1 overexpressing cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Immobilon®-P PVDF-Membran, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.