Merck
Early Corneal Wound Healing Response After Small Incision Lenticule Extraction.
Cornea (2019-08-23)
Yuan Sun, Ting Zhang, Manli Liu, Yugui Zhou, Shengbei Weng, Xiaonan Yang, Quan Liu
ZUSAMMENFASSUNG
To evaluate the corneal wound healing response after small incision lenticule extraction surgery. Small incision lenticule extraction was performed in both eyes of 12 New Zealand White rabbits. The refractive spherical correction was set at -6.00 D. Two animals were analyzed at each time point (1 hour, 4 hours, 1 day, 3 days, 7 days, and 28 days). The corneas were evaluated using slit-lamp and in vivo confocal microscopy. After euthanatization, the corneal tissues were subjected to light microscopy, transferase 2'-Deoxyuridine 5'-Triphosphate (dUTP) nick end labeling assay, and immunofluorescence microscopy (CD11b, fibronectin, tenascin, alpha-smooth muscle actin [α-SMA]). The corneas did not show any opacity at any time point except at the side-cut incision. By contrast, there was obvious scar tissue at the side-cut incision. Scattered, hyperreflective spots were seen by confocal microscopy from 1 hour postoperatively. Transferase dUTP nick end labeling-positive keratocytes were abundant near the femtosecond laser incision area at 1 hour and reached a peak at 4 hours postoperatively and then decreased. Inflammatory cells migrated from the incision into the central cornea, and this process began 1 hour after surgery and peaked at 7 days. Extracellular matrix components were deposited at the beginning of day 1 postoperatively, and the distribution pattern differed between the central cornea and the incision site. α-SMA-positive myofibroblasts were only detected at the side-cut incision. The scar tissue response in the peripheral cornea is related to the epithelium debridement. Inflammatory cells begin to be recruited by 1 hour after surgery. Therefore, it is necessary to implement antiinflammation interventions at a very early stage.