Merck
  • Inhibition of visfatin/NAMPT affects ovarian proliferation, apoptosis, and steroidogenesis in pre-pubertal mice ovary.
Inhibition of visfatin/NAMPT affects ovarian proliferation, apoptosis, and steroidogenesis in pre-pubertal mice ovary.
The Journal of steroid biochemistry and molecular biology (2020-09-29)
Lalrawngbawli Annie, Guruswami Gurusubramanian, Vikas Kumar Roy
ZUSAMMENFASSUNG
Pubertal ovarian function might be dependent on the factors present in the pre-pubertal stages. Visfatin regulates ovarian steroidogenesis in adult. To date, no study has investigated the role of visfatin either in pre-pubertal or pubertal mice ovary. Thus, we investigated the role of visfatin in pre-pubertal mice ovary in relation to steroidogenesis and proliferation and apoptosis in vitro by inhibiting the endogenous visfatin by a specific inhibitor, FK866. Inhibition of visfatin increased the estrogen secretion and also up-regulated the expression of CYP11A1, 17βHSD and CYP19A1 in mice ovary. Furthermore, active caspase3 was up-regulated along with the down-regulation of BAX and BCL2 in the pre-pubertal ovary after visfatin inhibition. The expression of GCNA, PCNA, and BrdU labeling was also decreased by FK866 treatment. These results suggest that visfatin inhibits steroidogenesis, increases proliferation, and suppresses apoptosis in the pre-pubertal mice ovary. So, visfatin is a new regulator of ovary function in pre-pubertal mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
FK866 hydrochloride hydrate, ≥98% (HPLC)