Merck
AMP-activated protein kinase is essential for survival in chronic hypoxia.
Biochemical and biophysical research communications (2008-03-25)
Darrell R Borger, L Cristina Gavrilescu, Maria C Bucur, Mircea Ivan, James A Decaprio
ZUSAMMENFASSUNG
This study was undertaken to interrogate cancer cell survival during long-term hypoxic stress. Two systems with relevance to carcinogenesis were employed: Fully transformed BJ cells and a renal carcinoma cell line (786-0). The dynamic of AMPK activity was consistent with a prosurvival role during chronic hypoxia. This was further supported by the effects of AMPK agonists and antagonists (AICAR and compound C). Expression of a dominant-negative AMPK alpha resulted in a decreased ATP level and significantly compromised survival in hypoxia. Dose-dependent prosurvival effects of rapamycin were consistent with mTOR inhibition being a critical downstream mediator of AMPK in persistent low oxygen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination