- Diffusive uptake rates of volatile organic compounds on standard ATD tubes for environmental and workplace applications
Diffusive uptake rates of volatile organic compounds on standard ATD tubes for environmental and workplace applications
Chemosphere (2017)
Jia C and Fu X