Merck
MC4R agonism promotes durable weight loss in patients with leptin receptor deficiency.
Nature medicine (2018-05-08)
Karine Clément, Heike Biebermann, I Sadaf Farooqi, Lex Van der Ploeg, Barbara Wolters, Christine Poitou, Lia Puder, Fred Fiedorek, Keith Gottesdiener, Gunnar Kleinau, Nicolas Heyder, Patrick Scheerer, Ulrike Blume-Peytavi, Irina Jahnke, Shubh Sharma, Jacek Mokrosinski, Susanna Wiegand, Anne Müller, Katja Weiß, Knut Mai, Joachim Spranger, Annette Grüters, Oliver Blankenstein, Heiko Krude, Peter Kühnen
ZUSAMMENFASSUNG
Genetic defects underlying the melanocortin-4 receptor (MC4R) signaling pathway lead to severe obesity. Three severely obese LEPR-deficient individuals were administered the MC4R agonist setmelanotide, resulting in substantial and durable reductions in hyperphagia and body weight over an observation period of 45-61 weeks. Compared to formerly developed and tested MC4R agonists, setmelanotide has the unique capability of activating nuclear factor of activated T cell (NFAT) signaling and restoring function of this signaling pathway for selected MC4R variants. Our data demonstrate the potency of setmelanotide in treatment of individuals with diverse MC4R-related pathway deficiencies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LY2112688 trifluoroacetate, ≥95% (HPLC)